missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FHL2 Protein
A cDNA sequence encoding the FHL2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11879-25ug
Additional Details : Weight : 0.01000kg
Specifications
FHL2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTERFDC HHCNESLFGK KYILREESPY CVVCFETLFA NTCEECGKPI GCDCKDLSYK DRHWHEACFH CSQCRNSLVD KPFAAKEDQL LCTDCYSNEY SSKCQECKKT IMPGTRKMEY KGSSWHETCF ICHRCQQPIG TKSFIPKDNQ NFCVPCYEKQ HAMQCVQCKK PITTGGVTYR EQPWHKECFV CTACRKQLSG QRFTARDDFA YCLNCFCDLY AKKCAGCTNP ISGLGGTKYI SFEERQWHND CFNCKKCSLS LVGRGFLTER DDILCPDCGK DI | |
Greater than 90.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FHL2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |