missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FLT1 D3 Protein
A cDNA sequence encoding the FLT1 D3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10612-10ug
Additional Details : Weight : 0.01000kg
Specifications
2321 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The activity of FLT1D1-3 was determined by its ability to inhibit the VEGF-165-induced proliferation of HUVE cells. | |
Human | |
Untagged | |
FLT1 D1-3 was lyophilized from a concentrated (1 mg/ml) sterile solution containing 1xPBS. |
FLT1 D3 Protein | |
Research Use Only | |
FLT1 | |
Recombinant Protein | |
Baculovirus | |
SKLKDPELSLKGTQHIMQAGQTLHLQCRGEAAHKWSLPEMVSKESERLSI TKSACGRNGKQFCSTLTLNTAQANHTGFYSCKYLAVPTSKKKETESAIYI FISDTGRPFVEMYSEIPEIIHMTEGRELVIPCRVTSPNITVTLKKFPLDT LIPDGKRIIWDSRKGFIISNATYKEIGLLTCEATVNGHLYKTNYLTHRQT NTIIDVQISTPRPVKLLRGHTLVLNCTATTPLNTRVQMTWSYPDEKNKRA SVRRRIDQSNSHANIFYSVLTIDKMQNKDKGLYTCRVRSGPSFKSVNTSV HIYDKAFITVKHRKQQVLETVAGKRSY | |
Greater than 90.0% as determined by SDS-PAGE. |