missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FLT4 Fc Protein
A cDNA sequence encoding the FLT4 Fc was constructed and used to recombinantly synthesize the protein.
131.00 € - 7250.00 €
Specifications
Name | FLT4 Fc Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Measured by its ability to bind recombinant rat VEGF-C in a functional solid phase binding assay. Immobilised recombinant human VEGFR-3/FLT-4 at 5 microg/ml can bind recombinant rat VEGF-C in a linear range of 8-500 ng/ml. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15904438
|
enQuireBio™
QP10619-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984428
|
enQuireBio™
QP10619-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994428
|
enQuireBio™
QP10619-1MG |
1 mg |
7250.00 €
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FLT4 Fc Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
MQRGAALCLR LWLCLGLLDG LVSGYSMTPP TLNITEESHV IDTGDSLSIS CRGQHPLEWA WPGAQEAPAT GDKDSEDTGV VRDCEGTDAR PYCKVLLLHE VHANDTGSYV CYYKYIKARI EGTTAASSYV FVRDFEQPFI NKPDTLLVNR KDAMWVPCLV SIPGLNVTLR SQSSVLWPDG QEVVWDDRRG MLVSTPLLHD ALYLQCETTW GDQDFLSNPF LVHITGNELY DIQLLPRKSL ELLVGEKLVL NCTVWAEFNS GVTFDWDYPG KQAERGKWVP ERRSQQTHTE LSSILTIHNV SQHDLGSYVC KANNGIQRFR ESTEVIVHEN PFISVEWLKG PILEATAGDE LVKLPVKLAA YPPPEFQWYK DGKALSGRHS PHALVLKEVT EASTGTYTLA LWNSAAGLRR NISLELVVNV PPQIHEKEAS SPSIYSRHSR QALTCTAYGV PLPLSIQWHW RPWTPCKMFA QRSLRRRQQQ DLMPQCRDWR AVTTQDAVNP IESLDTWTEF VEGKNKTVSK LVIQNANVSA MYKCVVSNKV GQDERLIYFY VTTIPDGFTI ESKPSEELLE GQPVLLSCQA DSYKYEHLRW YRLNLSTLHD AHGNPLLLDC KNVHLFATPL AASLEEVAPG ARHATLSLSI PRVAPEHEGH YVCEVQDRRS HDKHCHKKYL SVQALEAPRL TQNLTDLLVN VSDSLEMQCL VAGAHAPSIV WYKDERLLEE KSGVDLADSN QKLSIQRVRE EDAGRYLCSV CNAKGCVNSS ASVAVEGSED KGSMEIVILV GTGVIAVFFW VLLLLIFCNM RRPAHADIKT GYLSIIMDPG EVPLEEQCEY LSYDASQWEF PRERLHLGRV LGYGAFGKVV EASAFGIHKG SSCDTVAVKM LKEGATASEH RALMSELKIL IHIGNHLNVV | |
Greater than 90.0% as determined by(a)Analysis by RP-HPLC. (b)Analysis by SDS-PAGE. |
Research Use Only | |
Measured by its ability to bind recombinant rat VEGF-C in a functional solid phase binding assay. Immobilised recombinant human VEGFR-3/FLT-4 at 5 microg/ml can bind recombinant rat VEGF-C in a linear range of 8-500 ng/ml. | |
Human | |
Untagged | |
FLT4 Fc Chimera was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives. |