missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Forkhead box protein M1 Protein

A DNA sequence encoding the Homo sapiens (Human) Forkhead box protein M1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6054-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989649

  • 2720.00 € / 1mg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001230017.1
Forkhead box protein M1 Protein
Research Use Only
FOXM1
Recombinant Protein
E. coli
ERPPYSYMAM IQFAINSTER KRMTLKDIYT WIEDHFPYFK HIAKPGWKNS IRHNLSLHDM FVRETSANGK VSFWTIHPSA NRYLTLDQVF KPL
Greater than 80% as determined by SDS-PAGE.
2305
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers