missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fragile histidine triad / FHIT Protein
A DNA sequence encoding the Homo sapiens (Human) Fragile histidine triad / FHIT, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP1078-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001159715.1 | |
Fragile histidine triad / FHIT Protein | |
Research Use Only | |
FHIT | |
Recombinant Protein | |
E. coli | |
SFRFGQHLIK PSVVFLKTEL SFALVNRKPV VPGHVLVCPL RPVERFHDLR PDEVADLFQT TQRVGTVVEK HFHGTSLTFS MQDGPEAGQT VKHVHVHVLP RKAGDFHRND SIYEELQKHD KEDFPASWRS EEEMAAEAAA LRVYFQ | |
Greater than 90% as determined by SDS-PAGE. |
2272 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |