missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FTCD Protein
A cDNA sequence encoding the FTCD was constructed and used to recombinantly synthesize the protein.
131.00 € - 4785.00 €
Specifications
Gene ID (Entrez) | 10841 |
---|---|
Name | FTCD Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | FTCD |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968758
|
enQuireBio™
QP11906-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958758
|
enQuireBio™
QP11906-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948758
|
enQuireBio™
QP11906-1MG |
1 mg |
4785.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
10841 | |
Research Use Only | |
FTCD | |
Human | |
His | |
FTCD is supplied in 16mM HEPES buffer pH 7.6, 240mM sodium chloride, and 20% glycerol. |
FTCD Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MSQLVECVPN FSEGKNQEVI DAISGAITQT PGCVLLDVDA GPSTNRTVYT FVGPPECVVE GALNAARVAS RLIDMSRHQG EHPRMGALDV CPFIPVRGVS VDECVLCAQA FGQRLAEELD VPVYLYGEAA RMDSRRTLPA IRAGEYEALP KKLQQADWAP DFGPSSFVPS WGATATGARK FLIAFNINLL GTKEQAHRIA LNLREQGRGK DQPGRLKKVQ GIGWYLDEKN LAQVSTNLLD FEVTALHTVY EETCREAQEL SLPVVGSQLV GLVPLKALLD AAAFYCEKEN LFILEEEQRI RLVVSRLGLD SLCPFSPKER IIEYLVPERG PERGLGSKSL RAFVGEVGAR SAAPGGGSVA AAAAAMGAAL GSMVGLMTYG RRQFQSLDTT MRRLIPPFRE ASAKLTTLVD ADAEAFTAYL EAMRLPKNTP EEKDRRTAAL QEGLRRAVSV PLTLAETVAS LWPALQELAR CGNLACRSDL QVAAKALEMG VFGAYFNVLI NLRDITDEAF KDQIHHRVSS LLQEAKTQAA LVLDCLETRQ E | |
Greater than 95% as determined by SDS-PAGE. |