missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FTSJ1 Protein
A DNA sequence encoding the Homo sapiens (Human) FTSJ1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001269086.1 |
---|---|
Gene ID (Entrez) | 24140 |
Name | FTSJ1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15909679
|
enQuireBio™
QP6058-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929679
|
enQuireBio™
QP6058-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919679
|
enQuireBio™
QP6058-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15939679
|
enQuireBio™
QP6058-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15949679
|
enQuireBio™
QP6058-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15959679
|
enQuireBio™
QP6058-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001269086.1 | |
FTSJ1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
24140 | |
Research Use Only | |
FTSJ1 | |
Recombinant Protein | |
E. coli | |
MGRTSKDKRD VYYRLAKENG WRARSAFKLL QLDKEFQLFQ GVTRAVDLCA APGSWSQVLS QKIGGQGSGH VVAVDLQAMA PLPGVVQIQG DITQLSTAKE IIQHFKGCPA DLVVCDGAPD VTGLHDVDEY MQAQLLLAAL NIATHVLKPG GCFVAKIFRG RDVTLLYSQL QVFFSSVLCA KPRSSRNSSI EAFAVCQGYD PPEGFIPDLS KPLLDHSYDP DFNQLDGPTR IIVPFVTCGD LSSYDSDRSY PLDLEGGSEY KYTPPTQPPI SPPYQEACTL KRKGQLAKEI RPQDCPISRV DTFPQPLAAP QCHTLLAPEM EDNEMSCSP | |
Greater than 90% as determined by SDS-PAGE. |