missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human G-CSF / CSF3 Protein
A DNA sequence encoding the Homo sapiens (Human) G-CSF / CSF3, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_000750.1 |
---|---|
Gene ID (Entrez) | 1440 |
Name | G-CSF / CSF3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015201
|
enQuireBio™
QP8545-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16035201
|
enQuireBio™
QP8545-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16025201
|
enQuireBio™
QP8545-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045201
|
enQuireBio™
QP8545-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055201
|
enQuireBio™
QP8545-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065201
|
enQuireBio™
QP8545-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000750.1 | |
G-CSF / CSF3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1440 | |
Research Use Only | |
CSF3 | |
Recombinant Protein | |
E. coli | |
VQEATPLGPA SSLPQSFLLK CLEQVRKIQG DGAALQEKLV SECATYKLCH PEELVLLGHS LGIPWAPLSS CPSQALQLAG CLSQLHSGLF LYQGLLQALE GISPELGPTL DTLQLDVADF ATTIWQQMEE LGMAPALQPT QGAMPAFASA FQRRAGGVLV ASHLQSFLEV SYRV | |
Greater than 90% as determined by SDS-PAGE. |