missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GAD1 iso1 Protein
A cDNA sequence encoding the GAD1 iso1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 2571 |
---|---|
Name | GAD1 iso1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | GAD1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15928798
|
enQuireBio™
QP11918-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918798
|
enQuireBio™
QP11918-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15908798
|
enQuireBio™
QP11918-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
2571 | |
Research Use Only | |
GAD1 | |
Human | |
His | |
GAD1 iso1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
GAD1 iso1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASSTPS SSATSSNAGA DPNTTNLRPT TYDTWCGVAH GCTRKLGLKI CGFLQRTNSL EEKSRLVSAF KERQSSKNLL SCENSDRDAR FRRTETDFSN LFARDLLPAK NGEEQTVQFL LEVVDILLNY VRKTFDRSTK VLDFHHPHQL LEGMEGFNLE LSDHPESLEQ ILVDCRDTLK YGVRTGHPRF FNQLSTGLDI IGLAGEWLTS TANTNMFTYE IAPVFVLMEQ ITLKKMREIV GWSSKDGDGI FSPGGAISNM YSIMAARYKY FPEVKTKGMA AVPKLVLFTS EQSHYSIKKA GAALGFGTDN VILIKCNERG KIIPADFEAK ILEAKQKGYV PFYVNATAGT TVYGAFDPIQ EIADICEKYN LWLHVDAAWG GGLLMSRKHR HKLNGIERAN SVTWNPHKMM GVLLQCSAIL VKEKGILQGC NQMCAGYLFQ PDKQYDVSYD TGDKAIQCGR HVDIFKFWLM WKAKGTVGFE NQINKCLELA EYLYAKIKNR EEFEMVFNGE PEHTNVCFWY IPQSLRGVPD SPQRREKLHK VAPKIKALMM ESGTTMVGYQ PQGDKANFFR MVISNPAATQ SDIDFLIEEI ERLGQDL | |
Greater than 80% as determined by SDS-PAGE. |