missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Galactose-1-phosphate uridylyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) Galactose-1-phosphate uridylyltransferase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6067-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000146.2 | |
Galactose-1-phosphate uridylyltransferase Protein | |
Research Use Only | |
GALT | |
Recombinant Protein | |
E. coli | |
MSRSGTDPQQ RQQASEADAA AATFRANDHQ HIRYNPLQDE WVLVSAHRMK RPWQGQVEPQ LLKTVPRHDP LNPLCPGAIR ANGEVNPQYD STFLFDNDFP ALQPDAPSPG PSDHPLFQAK SARGVCKVMC FHPWSDVTLP LMSVPEIRAV VDAWASVTEE LGAQYPWVQI FENKGAMMGC SNPHPHCQVW ASSFLPDIAQ REERSQQAYK SQHGEPLLME YSRQELLRKE RLVLTSEHWL VLVPFWATWP YQTLLLPRRH VRRLPELTPA ERDDLASIMK KLLTKYDNLF ETSFPYSMGW HGAPTGSEAG ANWNHWQLHA HYYPPLLRSA TVRKFMVGYE MLAQAQRDLT PEQAAERLRA LPEVHYHLGQ KDRETATIA | |
Greater than 90% as determined by SDS-PAGE. |
2592 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |