missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GALK1 Protein

A cDNA sequence encoding the GALK1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11927-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988818

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

GALK1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GALK1 Human solution containing 20mM Tris pH 8.0, and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAALRQPQVA ELLAEARRAF REEFGAEPEL AVSAPGRVNL IGEHTDYNQG LVLPMALELM TVLVGSPRKD GLVSLLTTSE GADEPQRLQF PLPTAQRSLE PGTPRWANYV KGVIQYYPAA PLPGFSAVVV SSVPLGGGLS SSASLEVATY TFLQQLCPDS GTIAARAQVC QQAEHSFAGM PCGIMDQFIS LMGQKGHALL IDCRSLETSL VPLSDPKLAV LITNSNVRHS LASSEYPVRR RQCEEVARAL GKESLREVQL EELEAARDLV SKEGFRRARH VVGEIRRTAQ AAAALRRGDY RAFGRLMVES HRSLRDDYEV SCPELDQLVE AALAVPGVYG SRMTGGGFGG CTVTLLEASA APHAMRHIQE HYGGTATFYL SQAADGAKVL CL
Videos
SDS
Documents
Certificates