missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GFPT1 Protein
A cDNA sequence encoding the GFPT1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | GFPT1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968918
|
enQuireBio™
QP11956-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958918
|
enQuireBio™
QP11956-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948918
|
enQuireBio™
QP11956-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GFPT1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GFPT1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 50% glycerol, 2mM DTT and 2mM EDTA. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQQIMKGN FSSFMQKEIF EQPESVVNTM RGRVNFDDYT VNLGGLKDHI KEIQRCRRLI LIACGTSYHA GVATRQVLEE LTELPVMVEL ASDFLDRNTP VFRDDVCFFL SQSGETADTL MGLRYCKERG ALTVGITNTV GSSISRETDC GVHINAGPEI GVASTKAYTS QFVSLVMFAL MMCDDRISMQ ERRKEIMLGL KRLPDLIKEV LSMDDEIQKL ATELYHQKSV LIMGRGYHYA TCLEGALKIK EITYMHSEGI LAGELKHGPL ALVDKLMPVI MIIMRDHTYA KCQNALQQVV ARQGRPVVIC DKEDTETIKN TKRTIKVPHS VDCLQGILSV IPLQLLAFHL AVLRGYDVDF PRNLAKSVTV E | |
Greater than 95.0% as determined by SDS-PAGE. |