missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GHRH Protein
A cDNA sequence encoding the GHRH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10643-250ug
Additional Details : Weight : 0.01000kg
Specifications
GHRH Protein | |
Research Use Only | |
GHRH increases plasma growth hormone concentrations by directly stimulating the anterior pituitary gland to release natural human growth hormone. | |
Human | |
Untagged | |
The GHRH peptide (1 mg/ml) was lyophilized after extensive dialyses against 1.7 mg sodium phosphate buffer (0.1 mg sodium phosphate monobasic and 1.6 mg sodium phosphate dibasic). |
250 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The free base of sermorelin has the empirical formula C149H246N 44O42STyr-Ala-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-NH2 | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |