missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GKN2 Protein

A cDNA sequence encoding the GKN2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11978-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958988

  • 131.00 € / 2µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

GKN2 Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASYEVFNIISPS NNGGNVQETV TIDNEKNTAI INIHAGSCSS TTIFDYKHGY IASRVLSRRA CFILKMDHQN IPPLNNLQWY IYEKQALDNM FSSKYTWVKY NPLESLIKDV DWFLLGSPIE KLCKHIPLYK GEVVENTHNV GAGGCAKAGL LGILGISICA DIHV
Greater than 90.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GKN2 filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4.0.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers