missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GKN3P Protein

A cDNA sequence encoding the GKN3P was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11979-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988988

  • 131.00 € / 2µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GKN3P Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASMMNIRFNHPL YGSFGTQIIH IGAFQGMVSI RDNNIFSEWD GILDYKNALL VAKVFNKMAC VLARMDKAVF PSLDDISKAL DKQAFKYYPS TRGLTYTVLP SWVKNLAQYG KPIKNMCRDD PTYFAQQQKE GTALAIDSNS CFEIQLLSFM GLFICGETPG L
Greater than 95.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GKN3P filtered (0.4microm) solution at a concentration of 0.3 mg/ml in 0.03M acetate buffer, pH 4.0 and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers