missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GLIPR2 Protein
A cDNA sequence encoding the GLIPR2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11986-5ug
Additional Details : Weight : 0.01000kg
Specifications
GLIPR2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGKSASKQFH NEVLKAHNEY RQKHGVPPLK LCKNLNREAQ QYSEALASTR ILKHSPESSR GQCGENLAWA SYDQTGKEVA DRWYSEIKNY NFQQPGFTSG TGHFTAMVWK NTKKMGVGKA SASDGSSFVV ARYFPAGNVV NEGFFEENVL PPKK | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GLIPR2 1 mg/ml protein contains 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M DTT and 10% Glycerol. |