missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GLP-1R / GLP1R Protein
A DNA sequence encoding the Homo sapiens (Human) GLP-1R / GLP1R, was expressed in the hosts and tags indicated.
665.00 € - 1850.00 €
Specifications
Accession Number | NP_002053.3 |
---|---|
Gene ID (Entrez) | 2740 |
Name | GLP-1R / GLP1R Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16050740
|
enQuireBio™
QP6090-MA-20UG |
20 μg |
665.00 €
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040740
|
enQuireBio™
QP6090-MA-100UG |
100 μg |
1850.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060740
|
enQuireBio™
QP6090-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002053.3 | |
GLP-1R / GLP1R Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2740 | |
Research Use Only | |
Glp1r | |
Recombinant Protein | |
Mammal | |
RPQGATVSLW ETVQKWREYR RQCQRSLTED PPPATDLFCN RTFDEYACWP DGEPGSFVNV SCPWYLPWAS SVPQGHVYRF CTAEGLWLQK DNSSLPWRDL SECEESKRGE RSSPEEQLLF LY | |
Greater than 98.9% as determined by SDS-PAGE. |