missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GMFG Protein
A cDNA sequence encoding the GMFG was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12006-5ug
Additional Details : Weight : 0.01000kg
Specifications
GMFG Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSDSLVVCEV DPELTEKLRK FRFRKETDNA AIIMKVDKDR QMVVLEEEFQ NISPEELKME LPERQPRFVV YSYKYVHDDG RVSYPLCFIF SSPVGCKPEQQMMYAGSKNR LVQTAELTKV FEIRTTDDLT EAWLQEKLSF FR | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The GMF-gamma protein contains 20mM Tris-HCl pH 8.0, 1mM DTT, 1mM EDTA and 10% Glycerol. |