missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GMPS Protein
A cDNA sequence encoding the GMPS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12008-10ug
Additional Details : Weight : 0.01000kg
Specifications
GMPS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMALCNG DSKLENAGGD LKDGHHHYEG AVVILDAGAQ YGKVIDRRVR ELFVQSEIFP LETPAFAIKE QGFRAIIISG GPNSVYAEDA PWFDPAIFTI GKPVLGICYG MQMMNKVFGG TVHKKSVRED GVFNISVDNT CSLFRGLQKE EVVLLTHGDS VDKVADGFKV VARSGNIVAG IANESKKLYG AQFHPEVGLT ENGKVILKNF LYDIAGCSGT FTVQNRELEC IREIKERVGT SKVLVLLSGG VDSTVCTALL NRALNQEQVI AVHIDNGFMR KRESQSVEEA LKKLGIQVKV INAAHSFYNG TTTLPISDED RTPRKRISKT LNMTTSPEEK RKIIGDTFVK IANEVIGEMN LKPEEVFLAQ GTLRPDLIES ASLVASGKAE LIKTHHNDTE LIRKLREEGK VIEPLKDFHK DEVRILGREL GLPEELVSRH PFPGPGLAIR VICAEEPYIC KDFPETNNIL KIVADFSASV KKPHTLLQRV KACTTEEDQE KLMQITSLHS LNAFLLPIKT VGVQGDCRSY SYVCGISSKD EPDWESLIFL ARLIPRMCHN VNRVVYIFGP PVKEPPTDVT PTFLTTGVLS TLRQADFEAH NILRESGYAG KISQMPVILT PLHFDRDPLQ KQPSCQRSVV IRTFITSDFM TGIPATPGNE IPVEVVLKMV TEIKKIPGIS RIMYDLTSKP PGTTEWE | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GMPS solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol and 0.1M NaCl. |