missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNAQ Protein
A cDNA sequence encoding the GNAQ was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | GNAQ Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959088
|
enQuireBio™
QP12011-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949088
|
enQuireBio™
QP12011-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939088
|
enQuireBio™
QP12011-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GNAQ Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GNAQ protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTLESIM ACCLSEEAKE ARRINDEIER QLRRDKRDAR RELKLLLLGT GESGKSTFIK QMRIIHGSGY SDEDKRGFTK LVYQNIFTAM QAMIRAMDTL KIPYKYEHNK AHAQLVREVD VEKVSAFENP YVDAIKSLWN DPGIQECYDR RREYQLSDST KYYLNDLDRV ADPAYLPTQQ DVLRVRVPTT GIIEYPFDLQ SVIFRMVDVG GQRSERRKWI HCFENVTSIM FLVALSEYDQ VLVESDNENR MEESKALFRT IITYPWFQNS SVILFLNKKD LLEEKIMYSH LVDYFPEYDG PQRDAQAARE FILKMFVDLN PDSDKIIYSH FTCATDTENI RFVFAAVKDT ILQLNLKEYN LV | |
Greater than 90.0% as determined by SDS-PAGE. |