missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human gp100 / PMEL17 / SILV Protein
A DNA sequence encoding the Homo sapiens (Human) gp100 / PMEL17 / SILV, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6683-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186982.1 | |
gp100 / PMEL17 / SILV Protein | |
Research Use Only | |
SILV | |
Recombinant Protein | |
E. coli | |
KVPRNQDWLG VSRQLRTKAW NRQLYPEWTE AQRLDCWRGG QVSLKVSNDG PTLIGANASF SIALNFPGSQ KVLPDGQVIW VNNTIINGSQ VWGGQPVYPQ ETDDACIFPD GGPCPSGSWS QKRSFVYVWK TWGQYWQVLG GPVSGLSIGT GRAMLGTHTM EVTVYHRRGS RSYVPLAHSS SAFTITDQVP FSVSVSQLRA LDGGNKHFLR NQPLTFALQL HDPSGYLAEA DLSYTWDFGD SSGTLISRAL VVTHTYLEPG PVTAQVVLQA AIPLTSCGSS PVPGTTDGHR PTAEAPNTTA GQVPTTEVVG TTPGQAPTAE PSGTTSVQVP TTEVISTAPV QMPTAESTGM TPEKVPVSEV MGTTLAEMST PEATGMTPAE VSIVVLSGTT AAQVTTTEWV ETTARELPIP EPEGPDASSI MSTESITGSL GPLLDGTATL RLV | |
Greater than 90% as determined by SDS-PAGE. |
6490 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |