missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GPC4 Protein

A cDNA sequence encoding the GPC4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12034-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989148

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

GPC4 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GPC4 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSSSLP SNVCNDERMA AGNGNEDDCW NGKGKSRYLF AVTGNGLANQ GNNPEVQVDT SKPDILILRQ IMALRVMTSK MKNAYNGNDV DFFDISDESS GEGSGSGCEY QQCPSEFDYN ATDHAGKSAN EKADS
Videos
SDS
Documents
Certificates