missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GPHB5 Protein
A cDNA sequence encoding the GPHB5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12039-2ug
Additional Details : Weight : 0.01000kg
Specifications
GPHB5 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GPHB5 filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4. |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASASSGNL RTFVGCAVRE FTFLAKKPGC RGLRITTDAC WGRCETWEKP ILEPPYIEAH HRVCTYNETK QVTVKLPNCA PGVDPFYTYP VAIRCDCGAC STATTECETI |