missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Growth Hormone Receptor / GHR / GHBP Protein

A DNA sequence encoding the Homo sapiens (Human) Growth Hormone Receptor / GHR / GHBP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6083-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989839

Quantity Price
1 434.28 € / 100µg
6 408.22 € / 100µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_000154.1
Growth Hormone Receptor / GHR / GHBP Protein
100 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
2690
Greater than 81.9% as determined by SDS-PAGE.
Research Use Only
GHR
Recombinant Protein
E. coli
FSGSEATAAI LSRAPWSLQS VNPGLKTNSS KEPKFTKCRS PERETFSCHW TDEVHHGTKN LGPIQLFYTR RNTQEWTQEW KECPDYVSAG ENSCYFNSSF TSIWIPYCIK LTSNGGTVDE KCFSVDEIVQ PDPPIALNWT LLNVSLTGIH ADIQVRWEAP RNADIQKGWM VLEYELQYKE VNETKWKMMD PILTTSVPVY SLKVDKEYEV RVRSKQRNSG NYGEFSEVLY VTLPQMSQFT CEEDFY
Videos
SDS
Documents
Certificates