missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GSTM2 Protein
A cDNA sequence encoding the GSTM2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10659-20ug
Additional Details : Weight : 0.01000kg
Specifications
GSTM2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPMTLGYWNI RGLAHSIRLL LEYTDSSYEE KKYTMGDAPD YDRSQWLNEK FKLGLDFPNL PYLIDGTHKI TQSNAILRYI ARKHNLCGES EKEQIREDIL ENQFMDSRMQ LAKLCYDPDF EKLKPEYLQA LPEMLKLYSQ FLGKQPWFLG DKITFVDFIA YDVLERNQVF EPSCLDAFPN LKDFISRFEG LEKISAYMKS SRFLPRPVFT KMAVWGNK | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
His | |
The GSTM2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol,0.1M NaCl and 1mM DTT. |