missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GTF2A2 Protein
A DNA sequence encoding the Homo sapiens (Human) GTF2A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6129-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001307858.1 | |
GTF2A2 Protein | |
Research Use Only | |
GTF2A2 | |
Recombinant Protein | |
E. coli | |
MAYQLYRNTT LGNSLQESLD ELIQSQQITP QLALQVLLQF DKAINAALAQ RVRNRVNFRG SLNTYRFCDN VWTFVLNDVE FREVTELIKV DKVKIVACDG KNTGSNTTE | |
Greater than 80% as determined by SDS-PAGE. |
2958 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |