missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HAAO Protein
A cDNA sequence encoding the HAAO was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12098-1mg
Specifications
HAAO Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMERRLG VRAWVKENRG SFQPPVCNKL MHQEQLKVMF IGGPNTRKDY HIEEGEEVFY QLEGDMVLRV LEQGKHRDVV IRQGEIFLLP ARVPHSPQRF ANTVGLVVER RRLETELDGL RYYVGDTMDV LFEKWFYCKD LGTQLAPIIQ EFFSSEQYRT GKPIPDQLLK EPPFPLSTRS IMEPMSLDAW LDSHHRELQA GTPLSLFGDT YETQVIAYGQ GSSEGLRQNV DVWLWQLEGS SVVTMGGRRL SLAPDDSLLV LAGTSYAWER TQGSVALSVT QDPACKKPLG | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HAAO protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |