missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Haptoglobin Protein
A cDNA sequence encoding the Haptoglobin was constructed and used to recombinantly synthesize the protein.
143.00 € - 2755.00 €
Specifications
Gene ID (Entrez) | 3240 |
---|---|
Name | Haptoglobin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | HP |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15949378
|
enQuireBio™
QP12103-200UG |
200 μg |
143.00 €
200µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939378
|
enQuireBio™
QP12103-1MG |
1 mg |
433.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929378
|
enQuireBio™
QP12103-10MG |
10 mg |
2755.00 €
10mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
3240 | |
Research Use Only | |
HP | |
Human | |
Untagged | |
Lyophilized with no additives. |
Haptoglobin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MSALGAVIAL LLWGQLFAVD SGNDVTDIAD DGCPKPPEIA HGYVEHSVRY QCKNYYKLRT EGDGVYTLND KKQWINKAVG DKLPECEADD GCPKPPEIAH GYVEHSVRYQ CKNYYKLRTE GDGVYTLNNE KQWINKAVGD KLPECEAVCG KPKNPANPVQ RILGGHLDAK GSFPWQAKMV SHHNLTTGAT LINEQWLLTT AKNLFLNHSE NATAKDIAPT LTLYVGKKQL VEIEKVVLHP NYSQVDIGLI KLKQKVSVNE RVMPICLPSK DYAEVGRVGY VSGWGRNANF KFTDHLKYVM LPVADQDQCI RHYEGSTVPE KKTPKSPVGV QPILNEHTFC AGMSKYQEDT CYGDAGSAFA VHDLEEDTWY ATGILSFDKS CAVAEYGVYV KVTSIQDWVQ KTIAEN | |
Greater than 95.0%. |