missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HBEGF / DTR Protein
Recombinant Protein
107.00 € - 1885.00 €
Specifications
Accession Number | NP_001936.1 |
---|---|
Gene ID (Entrez) | 1839 |
Name | HBEGF / DTR Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935889
|
enQuireBio™
QP5305-10UG |
10 μg |
107.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965889
|
enQuireBio™
QP5305-50UG |
50 μg |
252.00 €
50µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925889
|
enQuireBio™
QP5305-100UG |
100 μg |
433.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955889
|
enQuireBio™
QP5305-500UG |
500 μg |
1110.00 €
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945889
|
enQuireBio™
QP5305-1MG |
1 mg |
1885.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001936.1 | |
HBEGF / DTR Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
3T3 cell proliferation is significantly induced by less than 1 ng/ml Human HBEGF / DTR, (activity greater than 1.0 x 10^6 units/mg). | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
1839 | |
Research Use Only | |
HBEGF | |
Recombinant Protein | |
E. coli | |
MDLQEADLDL . LRVTLSSKP Q. ALATPNKE EH. GKRKKKG KGL. GKKRDP CLRK. YKDFC IHGEC. KYVK ELRAPS. CIC HPGYHGERCH GLSL | |
Greater than 95% as determined by SDS-PAGE. |