missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Hexokinase-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Hexokinase-1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_000179.2 |
---|---|
Gene ID (Entrez) | 3098 |
Name | Hexokinase-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051270
|
enQuireBio™
QP6165-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071270
|
enQuireBio™
QP6165-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061270
|
enQuireBio™
QP6165-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081270
|
enQuireBio™
QP6165-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091270
|
enQuireBio™
QP6165-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001280
|
enQuireBio™
QP6165-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000179.2 | |
Hexokinase-1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3098 | |
Research Use Only | |
HK1 | |
Recombinant Protein | |
E. coli | |
HFHLTKDMLL EVKKRMRAEM ELGLRKQTHN NAVVKMLPSF VRRTPDGTEN GDFLALDLGG TNFRVLLVKI RSGKKRTVEM HNKIYAIPIE IMQGTGEELF DHIVSCISDF LDYMGIKGPR MPLGFTFSFP CQQTSLDAGI LITWTKGFKA TDCVGHDVVT LLRDAIKRRE EFDLDVVAVV NDTVGTMMTC AYEEPTCEVG LIVGTGSNAC YMEEMKNVEM VEGDQGQMCI NMEWGAFGDN GCLDDIRTHY DRLVDEYSLN AGKQRYEKMI SGMYLGEIVR NILIDFTKKG FLFRGQISET LKTRGIFETK FLSQIESDRL ALLQVRAILQ QLGLNSTCDD SILVKTVCGV VSRRAAQLCG AGMAAVVDKI RENRGLDRLN VTVGVDGTLY KLHPHFSRIM HQTVKELSPK CNVSFLLSED GSGKGAALIT AVGVRLRTEA SS | |
Greater than 90% as determined by SDS-PAGE. |