missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histamine H1 receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Histamine H1 receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6181-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000852.1 | |
Histamine H1 receptor Protein | |
Research Use Only | |
HRH1 | |
Recombinant Protein | |
E. coli | |
AKIYKAVRQH CQHRELINRS LPSFSEIKLR PENPKGDAKK PGKESPWEVL KRKPKDAGGG SVLKSPSQTP KEMKSPVVFS QEDDREVDKL YCFPLDIVHM QAAAEGSSRD YVAVNRSHGQ LKTDEQGLNT HGASEISEDQ MLGDSQSFSR TDSDTTTETA PGKGKLRSGS NTGLDYIKFT WKRLRSHSRQ YVSGLHMNRE RKAAKQ | |
Greater than 90% as determined by SDS-PAGE. |
3269 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |