missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone H2A.V Protein

A DNA sequence encoding the Homo sapiens (Human) Histone H2A.V, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7750-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16000581

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_036544.1
Histone H2A.V Protein
Research Use Only
H2AFV
Recombinant Protein
E. coli
MAGGKAGKDS GKAKAKAVSR SQRAGLQFPV GRIHRHLKTR TTSHGRVGAT AAVYSAAILE YLTAEVLELA GNASKDLKVK RITPRHLQLA IRGDEELDSL IKATIAGGGV IPHIHKSLIG KKGQQKTA
Greater than 90% as determined by SDS-PAGE.
94239
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers