missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-DRB1 Protein
A cDNA sequence encoding the HLA-DRB1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12276-5ug
Additional Details : Weight : 0.01000kg
Specifications
HLA-DRB1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGDTRPRFLW QPKRECHFFN GTERVRFLDR YFYNQEESVR FDSDVGEFRA VTELGRPDAE YWNSQKDILE QARAAVDTYC RHNYGVVESF TVQRRVQPKV TVYPSKTQPL QHHNLLVCSV SGFYPGSIEV RWFLNGQEEK AGMVSTGLIQ NGDWTFQTLV MLETVPRSGE VYTCQVEHPS VTSPLTVEWR ARSESAQSK | |
Greater than 85% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HLA-DRB1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |