missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMGN1 Protein
A cDNA sequence encoding the HMGN1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12285-2ug
Additional Details : Weight : 0.01000kg
Specifications
HMGN1 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HMGN1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M NaCl, 0.1mM PMSF and 20% glycerol. |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPKRKVSSAE GAAKEEPKRR SARLSAKPPA KVEAKPKKAA AKDKSSDKKV QTKGKRGAKG KQAEVANQET KEDLPAENGE TKTEESPASD EAGEKEAKSD LEHHHHHH |