missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HNRNPAB Protein

A cDNA sequence encoding the HNRNPAB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12290-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989948

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

HNRNPAB Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HNRNPAB protein solution (0.25 mg/ml) containing 30% glycerol and 1mM DTT.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSEAGEE QPMETTGATE NGHEAVPEGE SPAGAGTGAA AGAGGATAAP PSGNQNGAEG DQINASKNEE DAGKMFVGGL SWDTSKKDLK DYFTKFGEVV DCTIKMDPNT GRSRGFGFIL FKDAASVEKV LDQKEHRLDG RVIDPKKAMA MKKDPVKKIF VGGLNPEATE EKIREYFGEF GEIEAIELPM DPKLNKRRGF VFITFKEEEP VKKVLEKKFH TVSGSKCEIK VAQPKEVYQQ QQYGSGGRGN RNRGNRGSGG GGGGGGQGST NYGKSQRRGG HQNNYKPY
Videos
SDS
Documents
Certificates