missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HNRNPC Protein
A cDNA sequence encoding the HNRNPC was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Name | HNRNPC Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15919958
|
enQuireBio™
QP12291-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939958
|
enQuireBio™
QP12291-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929958
|
enQuireBio™
QP12291-50UG |
50 μg |
1740.00 €
50µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HNRNPC Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HNRNPC protein solution (0.25 mg/ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 5mM DTT and 30% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MASNVTNKTD PRSMNSRVFI GNLNTLVVKK SDVEAIFSKY GKIVGCSVHK GFAFVQYVNE RNARAAVAGE DGRMIAGQVL DINLAAEPKV NRGKAGVKRS AAEMYGSSFD LDYDFQRDYY DRMYSYPARV PPPPPIARAV VPSKRQRVSG NTSRRGKSGF NSKSGQRGSS KSGKLKGDDL QAIKKELTQI KQKVDSLLEN LEKIEKEQSK QAVEMKNDKS EEEQSSSSVK KDETNVKMES EGGADDSAEE GDLLDDDDNE DRGDDQLELI KDDEKEAEEG EDDRDSANGE DDS | |
Greater than 85% as determined by SDS-PAGE. |