missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HOXB13 Protein

A cDNA sequence encoding the HOXB13 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12296-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989968

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

HOXB13 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HOXB13 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMEPGNYA TLDGAKDIEG LLGAGGGRNL VAHSPLTSHP AAPTLMPAVN YAPLDLPGSA EPPKQCHPCP GVPQGTSPAP VPYGYFGGGY YSCRVSRSSL KPCAQAATLA AYPAETPTAG EEYPSRPTEF AFYPGYPGTY QPMASYLDVS VVQTLGAPGE PRHDSLLPVD SYQSWALAGG WNSQMCCQGE QNPPGPFWKA AFADSSGQHP PDACAFRRGR KKRIPYSKGQ LRELEREYAA NKFITKDKRR KISAATSLSE RQITIWFQNR RVKEKKVLAK VKNSATP
Videos
SDS
Documents
Certificates