missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSBP1L1 Protein

A cDNA sequence encoding the HSBP1L1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12318-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990039

  • 203.00 € / 20µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HSBP1L1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMDVRGPE APGGRALRDA AENLFQELQE HFQALTATLN LRMEEMGNRI EDLQKNVNDL MVQAGIENSI KEQMLKT
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HSBP1L1 protein solution (1 mg/ml) containing Phosphate buffer saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers