missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSPB7 Protein
A cDNA sequence encoding the HSPB7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12334-1mg
Specifications
HSPB7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSHRTSSTFR AERSFHSSSS SSSSSTSSSA SRALPAQDPP MEKALSMFSD DFGSFMRPHS EPLAFPARPG GAGNIKTLGD AYEFAVDVRD FSPEDIIVTT SNNHIEVRAE KLAADGTVMN TFAHKCQLPE DVDPTSVTSA LREDGSLTIR ARRHPHTEHV QQTFRTEIKI | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HSPB7 Human solution containing 20mM Tris HCl pH 8.0, 0.2M NaCl, 2mM DTT and 50% glycerol. |