missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSPB8 Protein
A cDNA sequence encoding the HSPB8 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1825.00 €
Specifications
Gene ID (Entrez) | 26353 |
---|---|
Name | HSPB8 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | HSPB8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930099
|
enQuireBio™
QP12335-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920099
|
enQuireBio™
QP12335-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15910099
|
enQuireBio™
QP12335-100UG |
100 μg |
1825.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
26353 | |
Research Use Only | |
HSPB8 | |
Human | |
Untagged | |
The HSPB8 protein was lyophilized from a concentrated (1 mg/ml) solution containing 20mM Tris-acetate, pH 7.6, 10mM NaCl, 0.1mM EDTA, 0.1mM PMSF, 15mM b-ME. |
HSPB8 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MADGQMPFSC HYPSRLRRDP FRDSPLSSRL LDDGFGMDPF PDDLTASWPD WALPRLSSAW PGTLRSGMVP RGPTATARFG VPAEGRTPPP FPGEPWKVCV NVHSFKPEEL MVKTKDGYVE VSGKHEEKQQ EGGIVSKNFT KKIQLPAEVD PVTVFASLSP EGLLIIEAPQ VPPYSTFGES SFNNELPQDS QEVTCT | |
Greater than 95.0% as determined by SDS-PAGE. |