missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Hyaluronidase PH-20 Protein
A DNA sequence encoding the Homo sapiens (Human) Hyaluronidase PH-20, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001167515.1 |
---|---|
Gene ID (Entrez) | 6677 |
Name | Hyaluronidase PH-20 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035360
|
enQuireBio™
QP6719-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055360
|
enQuireBio™
QP6719-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045360
|
enQuireBio™
QP6719-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065360
|
enQuireBio™
QP6719-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075360
|
enQuireBio™
QP6719-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085360
|
enQuireBio™
QP6719-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001167515.1 | |
Hyaluronidase PH-20 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6677 | |
Research Use Only | |
SPAM1 | |
Recombinant Protein | |
E. coli | |
LNFRAPPVIP NVPFLWAWNA PSEFCLGKFD EPLDMSLFSF IGSPRINATG QGVTIFYVDR LGYYPYIDSI TGVTVNGGIP QKISLQDHLD KAKKDITFYM PVDNLGMAVI DWEEWRPTWA RNWKPKDVYK NRSIELVQQQ NVQLSLTEAT EKAKQEFEKA GKDFLVETIK LGKLLRPNHL WGYYLFPDCY NHHYKKPGYN GSCFNVEIKR NDDLSWLWNE STALYPSIYL NTQQSPVAAT LYVRNRVREA IRVSKIPDAK SPLPVFAYTR IVFTDQVLKF LSQDELVYTF GETVALGASG IVIWGTLSIM RSMKSCLLLD NYMETILNPY IINVTLAAKM CSQVLCQEQG VCIRKNWNSS DYLHLNPDNF AIQLEKGGKF TVRGKPTLED LEQFSEKFYC SCYSTLSCKE KADVKDTDAV DVCIADGVCI DAFLKPPMET EEPQIFYNAS PSTLS | |
Greater than 90% as determined by SDS-PAGE. |