missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IDO2 Protein

A DNA sequence encoding the Homo sapiens (Human) IDO2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6196-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051480

  • 518.00 € / 100µg
Estimated Shipment: 28-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Descripción

Descripción

Especificaciones

Especificaciones

NP_919270.2
IDO2 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
169355
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IDO2
Recombinant Protein
E. coli
MEPHRPNVKT AVPLSLESYH ISEEYGFLLP DSLKELPDHY RPWMEIANKL PQLIDAHQLQ AHVDKMPLLS CQFLKGHREQ RLAHLVLSFL TMGYVWQEGE AQPAEVLPRN LALPFVEVSR NLGLPPILVH SDLVLTNWTK KDPDGFLEIG NLETIISFPG GESLHGFILV TALVEKEAVP GIKALVQATN AILQPNQEAL LQALQRLRLS IQDITKTLGQ MHDYVDPDIF YAGIRIFLSG WKDNPAMPAG LMYEGVSQEP LKYSGGSAAQ STVLHAFDEF LGIRHSKESG DFLYRMRDYM PPSHKAFIED IHSAPSLRDY ILSSGQDHLL TAYNQCVQAL AELRSYHITM VTKYLITAAA KAKHGKPNHL PGPPQALKDR GTGGTAVMSF LKSVRDKTLE SILHPRG
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales