missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFN a 1a Protein
A cDNA sequence encoding the IFN a 1a was constructed and used to recombinantly synthesize the protein.
131.00 € - 1305.00 €
Specifications
Name | IFN a 1a Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The specific activity as determined in a viral resistance assay viral ressistance assay was found to be 100,000,000IU/ mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15984598
|
enQuireBio™
QP10672-20UG |
20 μg |
131.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964598
|
enQuireBio™
QP10672-100UG |
100 μg |
203.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974598
|
enQuireBio™
QP10672-1MG |
1 mg |
1305.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IFN a 1a Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
CDLPETHSLD NRRTLMLLAQ MSRISPSSCL MDRHDFGFPQ EEFDGNQFQK APAISVLHEL IQQIFNLFTT KDSSAAWDED LLDKFCTELY QQLNDLEACV MQEERVGETP LMNADSILAV KKYFRRITLY LTEKKYSPCA WEVVRAEIMR SLSLSTNLQE RLRRKE | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
The specific activity as determined in a viral resistance assay viral ressistance assay was found to be 100,000,000IU/ mg. | |
Human | |
Untagged | |
The IFN-a 1a protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |