missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFN a 2b Protein
A cDNA sequence encoding the IFN a 2b was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10674-EC-PEG-10ug
Additional Details : Weight : 0.01000kg
Specifications
IFN a 2b Protein | |
Research Use Only | |
The specific activity was determined by the dose-dependent cytotoxicity of the human TF-1 cell line (human erythroleukemic indicator cell line) and is typically 0.02-0.08ng/ml. | |
Human | |
20kD PEG | |
The IFN-a 2b was lyophilized from 1 mg/ml in 1xPBS. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MCDLPQTHSL GSRRTLMLLA QMRRISLFSC LKDRHDFGFP QEEFGNQFQK AETIPVLHEM IQQIFNLFST KDSSAAWDET LLDKFYTELY QQLNDLEACV IQGVGVTETP LMKEDSILAV RKYFQRITLY LKEKKYSPCA WEVVRAEIMR SFSLSTNLQE SLRSKE | |
Greater than 95% as obsereved by SDS-PAGE. |