missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFNA7 Protein
A cDNA sequence encoding the IFNA7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12375-10ug
Additional Details : Weight : 0.01000kg
Specifications
3444 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMCDLPQ THSLRNRRAL ILLAQMGRIS PFSCLKDRHE FRFPEEEFDG HQFQKTQAIS VLHEMIQQTF NLFSTEDSSA AWEQSLLEKF STELYQQLND LEACVIQEVG VEETPLMNED FILAVRKYFQ RITLYLMEKK YSPCAWEVVR AEIMRSFSFS TNLKKGLRRK D | |
Greater than 90% as determined by SDS-PAGE. |
IFNA7 Protein | |
Research Use Only | |
IFNA7 | |
Human | |
His | |
IFNA7 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |