missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFT20 Protein
A cDNA sequence encoding the IFT20 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12379-5ug
Additional Details : Weight : 0.01000kg
Specifications
IFT20 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAKDILGEAG LHFDELNKLR VLDPEVTQQT IELKEECKDF VDKIGQFQKI VGGLIELVDQ LAKEAENEKM KAIGARNLLK SIAKQREAQQ QQLQALIAEK KMQLERYRVE YEALCKVEAE QNEFIDQFIF QK | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The IFT20 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 30% glycerol. |