missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IGFBP 1 Protein

A cDNA sequence encoding the IGFBP 1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10683-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984638

  • 3401.86 € / 1mg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

IGFBP 1 Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Native Protein
APWQCAPCSA EKLALCPPVS ASCSEVTRSA GCGCCPMCAL PLGAACGVAT ARCARGLSCR ALPGEQQPLH ALTRGQGACV QESDASAPHA AEAGSPESPE STEITEEELL DNFHLMAPSE EDHSILWDAI STYDGSKALH VTNIKKWKEP CRIELYRVVE SLAKAQETSG EEISKFYLPN CNKNGFYHSR QCETSMDGEA GLCWCVYPWN GKRIPGSPEI RGDPNCQIYF NVQN
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
The ED50, as determined by the inhibition of rHuIGF-I-induced proliferation of human MCF-7 cells, is less than 4microg/ml.
Human
Untagged
IGFBP-1 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS.
Videos
SDS
Documents
Certificates