missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IgG receptor FcRn large subunit p51 Protein

A DNA sequence encoding the Homo sapiens (Human) IgG receptor FcRn large subunit p51, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6030-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989479

Quantity Price
1 1407.27 € / 100µg
3 1323.54 € / 100µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_001129491.1
IgG receptor FcRn large subunit p51 Protein
100 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
2217
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FCGRT
Recombinant Protein
Mammal
AESHLSLLYH LTAVSSPAPG TPAFWVSGWL GPQQYLSYNS LRGEAEPCGA WVWENQVSWY WEKETTDLRI KEKLFLEAFK ALGGKGPYTL QGLLGCELGP DNTSVPTAKF ALNGEEFMNF DLKQGTWGGD WPEALAISQR WQQQDKAANK ELTFLLFSCP HRLREHLERG RGNLEWKEPP SMRLKARPSS PGFSVLTCSA FSFYPPELQL RFLRNGLAAG TGQGDFGPNS DGSFHASSSL TVKSGDEHHY CCIVQHAGLA QPLRVELESP AKSS
Videos
SDS
Documents
Certificates