missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IHH Protein
Recombinant Protein
Brand: enQuireBio™ QP5308-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002172.2 | |
IHH Protein | |
Research Use Only | |
IHH | |
Recombinant Protein | |
E. coli | |
MIIGPGRVVG . SRRRPPRKL V. PLAYKQFS PN. VPEKTLG ASG. RYEGKI ARSS. ERFKE LTPNY. NPDI IFKDEE. NTG ADRLMTQRCK DRLNSLA. IS VMNQWPGV. K LRVTEGWDE. DGHHSEESLH . YEGRAVDIT T. SDRDRNKY GL. LARLAVE AGF. DWVYYE SKAH. VHCSV KSEHSAAAKT GG | |
Greater than 95% as determined by SDS-PAGE. |
3549 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human IHH induces alkaline phosphatase activity in CCL-226 cells (typical ED50 is ? 10 ug/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |