missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 27 Protein
A cDNA sequence encoding the IL 27 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10706-1ug
Additional Details : Weight : 0.01000kg
Specifications
246778 | |
1 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity was determined by the dose dependent inhibition of HIV replication in human CD4+ T cells and in human monocyte-derived macrophages and is typically 4-12ng/ml. | |
Human | |
Untagged | |
The IL-27 was lyophilized from 1 mg/ml in 1xPBS. |
IL 27 Protein | |
Research Use Only | |
IL27 | |
Recombinant Protein | |
Human (HEK293) | |
IL-27A (215aa):FPRPPGRPQL SLQELRREFT VSLHLARKLL AEVRGQAHRF AESHLPGVNL YLLPLGEQLP DVSLTFQAWR RLSDPERLCF ISTTLQPFHA LLGGLGTQGR WTNMERMQLW AMRLDLRDLQ RHLRFQVLAA GFNLPEEEEE EEEEEEEERK GLLPGALGSA LQGPAQVSWP QLLSTYRLLH SLELVLSRAV RELLLLSKAG HSVWPLGFPT LSPQP IL-27B (209aa):RKGPPAALTL PRVQCRASRY PIAVDCSWTL PPAPNSTSPV SFIATYRLGM AARGHSWPCL QQTPTSTSCT ITDVQLFSMA PYVLNVTAVH PWGSSSSFVP FITEHIIKPD PPEGVRLSPL AERQLQVQWE PPGSWPFPEI FSLKYWIRYK RQGAARFHRV GPIEATSFIL RAVRPRARYY VQVAAQDLTD YGELSDWSLP ATATMSLGK | |
Greater than 90% as obsereved by SDS-PAGE. |